Would you like to log in again?
VWR® Contour™ Standing Height Combo Base Cabinet
Supplier: VWR
VWR® Contour™
CFE-2456-L CWE-2456-A CFE-2458-L CFE-2456-A CWE-2458-A CWE-2458-L CWE-2456-L CWE-2436-A CFE-2458-12 CWE-2438-A CFE-2438-A CFE-2436-A CFE-2458-A
CFE-2456-AEA 2719.11 USD InStock
CFE-2456-A CFE-2458-A CWE-2458-L CFE-2456-L CFE-2458-L CFE-2458-12 CWE-2456-L CWE-2456-A CFE-2438-A CFE-2436-A CWE-2438-A CWE-2458-A CWE-2436-A
VWR® Contour™ Standing Height Combo Base Cabinet
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface (not included) creates an overall height of 91.4 cm (36"). All cupboard cabinets have one full-width adjustable shelf.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty