Would you like to log in again?
VWR® Contour™ Standing Height Sink Cabinets
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CWG-2004-L CFG-2008-A CFG-20452A CFG-2006-P CWG-20452A CFG-2005-A CFG-20458A CWG-2008-A CFG-2004-L CWG-2009-A CWG-2005-A CFG-2006-A CWG-2006-A CWG-20458A CFG-2009-A CWG-2004-A CFG-2004-A
CWG-2006-AEA 2160 USD InStock
CWG-2006-A CFG-2004-L CWG-2005-A CWG-2004-A CWG-2009-A CWG-2008-A CWG-2004-L CFG-2005-A CFG-2006-A CFG-2008-A CFG-2009-A CWG-20458A CFG-2004-A CFG-2006-P CFG-20458A CWG-20452A CFG-20452A
VWR® Contour™ Standing Height Sink Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). All cupboard cabinets have one full-width adjustable shelf. Sink cabinets with center bench end comes furnished with finished back panels that may be removed when not required. Finished back adds 2.54 cm (1") to depth, for an overall cabinet depth of 58.4 cm (23").These cabinet includes supports.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty