Would you like to log in again?
VWR® Contour™ Fume Hood Base Cabinets
Supplier: VWR
VWR®
G08C3520600300 CWG-2085-V CFG-2084-V CFG-2086-V CWG-2084-V CFG-2088-V CFG-2085-V CWG-2088-V CWG-2086-V
CFG-20860VEA 2102.3 USD InStock
CFG-20860V CFG-2086-V CFG-2085-V CWG-2088-V CFG-2084-V CWG-2086-V CWG-2085-V CWG-2084-V CFG-2088-V
VWR® Contour™ Fume Hood Base Cabinets
Furniture Storage Cabinets
VWR® Venturi fume hood base cabinet.
VWR®/VFH-06/19-6 Steel cabinets (35" high) standing height steel and wood fume hood base cabinets are 35" high and a nominal 20" deep. ADA height steel fume hood base cabinets are 32" high and 20" deep; ADA height wood fume hood base cabinets are 32½" high and nominal 20" deep. Lengths as shown.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty