Would you like to log in again?
VWR FUME HOOD ACID STORAGE BASE 35X60IN
Supplier: VWR International
VWR* Fume Hood, Acid Storage, Base Cabinet, Steel construction with one piece polyethethylene liner, Left Hand Hinged, Size: 35 in high X 60 in wide
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR FUME HOOD ACID STORAGE BASE 35X60IN CFG-2089-V Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty