Would you like to log in again?
VWR® Contour™ Standing Height Pump Storage Cabinets
Supplier: VWR
VWR®
CWG-2354-A CWG-2354-L CFG-2354-A CFG-2354-L
CWG-2354-LEA 5074.23 USD InStock
CWG-2354-L CFG-2354-A CFG-2354-L CWG-2354-A
VWR® Contour™ Standing Height Pump Storage Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • The interior is lined with 1" thick neoprene foam (for sound deadening)
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above
A 2.54 cm (1”) thick work surface creates an overall height of 91.4 cm (36"). Cupboard cabinets are supplied with one full-width adjustable shelf.
(Requires a 2" hole in the hood work-surface). Each cabinet is furnished with 120 VAC, 20 amp, duplex receptacle mounted on the inside cabinet back and the pilot lighted toggle switch mounted in the top front rail (Wiring is not included).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty