Would you like to log in again?
VWR® Contour™ ADA Height Base Cabinets, Fume Hood Cabinets
Supplier: VWR
VWR®
CWG-3086-A CWG-3084-A CWG-3085-A CFG-3088-A CFG-3084-A CWG-3084-L CWG-3088-A CFG-3086-A CFG-3084-L CFG-3085-A
CFG-3084-LEA 1483.18 USD InStock
CFG-3084-L CWG-3085-A CWG-3086-A CFG-3084-A CWG-3088-A CFG-3085-A CFG-3086-A CWG-3084-L CFG-3088-A CWG-3084-A
VWR® Contour™ ADA Height Base Cabinets, Fume Hood Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 83.8 cm (33"). All cupboard cabinets are supplied with one full-width adjustable shelf.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty