Would you like to log in again?
VWR® Contour™ Kemstrut Assemblies
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
K-0012-0A K-0011-0A CFK-0013-0A
CFK-0012-0AEA 103.07 USD InStock
CFK-0012-0A CFK-0011-0A CFK-0013-0A
VWR® Contour™ Kemstrut Assemblies
Furniture Casework
Adjustable-height floor-mounted support for the rear overhang of fume hood worktops.
  • Smooth, easy-to-maintain surfaces
  • Color: Light neutral
It can also be used as a worktop and/or a drainline and piping support in the plumbing space of 363/4x31" wall tables and 363/4x54" or 363/4x55" center tables.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty