Would you like to log in again?
VWR® Contour™ Standing Height Base Cabinet Sink Supports
Supplier: VWR
VWR®
CFK-MS60-A CFK-MS36-A CFK-MS58-A CFK-MS30-A CFK-MS52-A CFK-MS48-A CFK-MS24-A
CFK-MS30-AEA 99.8 USD InStock
CFK-MS30-A CFK-MS52-A CFK-MS60-A CFK-MS36-A CFK-MS24-A CFK-MS58-A CFK-MS48-A
VWR® Contour™ Standing Height Base Cabinet Sink Supports
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Sink supports are designed to fit with a wide variety of cabinet sizes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty