Would you like to log in again?
VWR® Contour™ Adjustable Slip Scribe with Slip-On Angle, Steel
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CWL-0329-37 CFL-0329-37 CFL-0329-84
CFL-0329-37EA 112.91 USD InStock
CFL-0329-37 CWL-0329-37 CFL-0329-84
VWR® Contour™ Adjustable Slip Scribe with Slip-On Angle, Steel
Furniture Casework
Steel adjustable slip scribes are designed the same as standard scribes with the addition of a slip-on angle at the wall edge to dress-up the edge of the scribe.
  • Baked, chemical-resistant, powder-coat finish
  • Light neutral color
The steel adjustable slip scribe is designed to be cut to width in the field to more easily fit the space between the wall and the end of the furniture assembly.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty