Would you like to log in again?
VWR® Contour™ Wallcase Intermediate Filler
Supplier: VWR
VWR®
CWL-0630-W CFL-0630-W
CWL-0630-WEA 174.05 USD InStock
CWL-0630-W CFL-0630-W
VWR® Contour™ Wallcase Intermediate Filler
Furniture Casework
Closes openings between two adjacent wall cabinets.
  • Steel cabinets are phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Light neutral color
Intermediate filler is smooth-surfaced and easy to maintain.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty