Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Contour™ Finished Back Panels, Standing Height

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR® Contour™
CWL-6036-F CFL-4230-F CFL-6036-F CFL-2430-F CWL-4836-F CFL-3636-F CFL-3030-F CWL-3636-F CFL-1836-F CFL-3630-F CWL-1830-F CFL-1830-F CFL-3036-F CWL-3630-F CFL-4236-F CWL-1836-F CWL-3036-F CWL-4236-F CFL-4836-F CWL-6030-F CFL-2436-F CWL-2436-F CWL-3030-F CFL-4830-F CFL-6030-F CWL-4230-F CWL-2430-F
CWL-4836-FEA 453 USD InStock
CWL-4836-F CWL-3036-F CWL-1830-F CWL-6036-F CWL-3030-F CWL-4230-F CFL-1830-F CFL-4236-F CWL-6030-F CFL-1836-F CFL-4230-F CFL-4836-F CWL-1836-F CWL-4236-F CFL-2430-F CFL-3636-F CWL-2436-F CFL-3630-F CFL-2436-F CWL-3630-F CWL-3636-F CFL-3030-F CFL-6030-F CFL-4830-F CFL-6036-F CFL-3036-F CWL-2430-F
VWR® Contour™ Finished Back Panels, Standing Height
Furniture Casework
Phosphate-coated with a baked, chemical-resistant, powder-coat finish.

  • Smooth, easy-to-maintain surfaces
  • Panel adds 2.5 cm (1") to back of base cabinet
  • lLight neutral color

For backs of base cabinets exposed to view. Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty