Would you like to log in again?
VWR® Wall Shelf Brackets, Fixed and Adjustable
Supplier: VWR
VWR®
CWM-0062-AD CWM-0061-AD CFM-0038-AD CWM-0038-AD CFM-0062-AD CFM-0060-AD CFM-0031-0A CWM-0039-AD CWM-0031-0A CWF-0170-04 CFM-0044-AD CFF-0170-04 CWM-0044-AD CFM-0061-AD CFM-0039-AD
CWM-0038-ADEA 80.37 USD InStock
CWM-0038-AD CWM-0039-AD CFF-0170-04 CFM-0060-AD CFM-0031-0A CFM-0062-AD CFM-0044-AD CFM-0061-AD CFM-0039-AD CWF-0170-04 CWM-0062-AD CFM-0038-AD CWM-0061-AD CWM-0031-0A CWM-0044-AD
VWR® Wall Shelf Brackets, Fixed and Adjustable
Furniture Shelves
Fixed and adjustable wall-mounted brackets.
  • Designed to suit a wide array of storage applications
  • Fixed brackets mount directly to the wall
  • Adjustable brackets are designed to mount to vertical wall supports (sold separately)
Fixed brackets features a flat top surface for simple shelf attachment. Adjustable brackets features a lipped top surface to provide increased security and stability when shelves are attached.
Ordering information: Fixed shelf brackets are supplied without mounting hardware. Adjustable brackets come with mounting hardware.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty