Would you like to log in again?
VWR® Tall Storage Units with Glazed Door
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CFS50841618-R CFS50841348-A CFS50841330-A CFS50841618-L CFS50841336-A CFS50841648-A CWS50841618-L CFS50841624-L CFS50842224-L CFS50842242-A CWS50842218-A CFS50841630-A CWS50842236-A CWS50842230-A CFS50842218-L CWS50841642-A CFS50841318-R CWS50841618-A CFS50841636-A CWS50841648-A CFS50842224-R CFS50841624-R CFS50841342-A CFS50841324-R CWS50841636-A CWS50842242-A CFS50842248-A CFS50841642-A CWS50842248-A CFS50842230-A CFS50842236-A CWS50842218-L CFS50841324-L CWS50841630-A CFS50842218-R CFS50841318-L
CWS50841642-AEA 6972.8 USD InStock
CWS50841642-A CFS50842236-A CWS50842236-A CFS50841648-A CFS50842230-A CFS50841324-R CFS50841330-A CWS50841618-L CFS50841624-L CWS50842230-A CFS50842224-R CFS50841624-R CWS50842218-L CFS50842224-L CFS50841348-A CWS50842218-A CFS50841630-A CFS50841324-L CWS50841648-A CFS50842248-A CWS50841630-A CFS50841636-A CWS50842242-A CFS50841318-R CWS50842248-A CFS50842242-A CFS50841342-A CFS50842218-R CFS50842218-L CFS50841618-L CFS50841336-A CFS50841618-R CFS50841318-L CWS50841618-A CWS50841636-A CFS50841642-A
VWR® Tall Storage Units with Glazed Door
Furniture Storage Cabinets
Full height storage cabinets are 84" high and 16 or 22" deep. Lengths as shown. They are furnished with five adjustable shelves.
  • Full height
  • Glazed doors
  • Multiple depths
The fully-welded, heavy-gauge, steel cabinets are designed to SEFA 8 performance standards and built to rigorous benchmarks.
Certifications: Meets SEFA 8 performance standards.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty