Would you like to log in again?
Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Supplier: Cambridge
Camco® Erado-Sol®
207-32 207-G 307-2 207-32 207-16 307-8 207-16 307-G 307-32 207-G 307-16
10853-876EA 117.73 USD InStock
CA10853-876 CACM207 10853-876 CA75833-978 CA75833-982 CA75833-980 CM207 CM207C CA75833-976 CA75833-984 CACM207C
Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Skincare Soaps and Skin Cleansers
Fast and effective stain remover for most medical, histological, and chemical stains including iodine, Wright stain, Lugol’s stain, and fresh blood
Removes stains from hands, plastic, glassware, and laboratory equipment. Simply rub in gently, rinse with water, and stain is removed in a matter of seconds. Stain remover is biodegradable and phosphate-free with a pH of 7–8.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty