Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Camco® Wright Stain Buffer, pH 7.15, Cambridge
Supplier: Cambridge
Camco®
402 402
CMB41363PK 211.02 USD InStock
CACMB41363 CMB41363
Camco® Wright Stain Buffer, pH 7.15, Cambridge
Buffers Microscopy Buffers
Each vial contains enough buffer to prepare one gallon of pH 7.15 buffer solution for use in the five-step staining process
The buffer is helpful with the development of the correct, clear nuclear color schemes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty