Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EVERHUTCH CORECART Two-Bay Procedure Cabinets, Kewaunee

Supplier: Kewaunee

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MCT200-02-S2B MCT201-01-G2A MCT201-01-G2C MCT201-02-G2D MCT201-02-S2A MCT201-01-G2D MCT201-02-G2B MCT201-02-G2C MCT201-01-C2A MCT201-02-G2A MCT200-01-S2B MCT201-01-S2A MCT201-01-G2B MCT201-02-C2A
CT20102C2AEA 11340 USD InStock
CT20102C2A CT20101C2A CT20102S2A CT20101S2A CT20001S2B CT20002S2B CT20102G2C CT20101G2B CT20101G2C CT20102G2D CT20101G2D CT20102G2A CT20101G2A CT20102G2B
EVERHUTCH CORECART Two-Bay Procedure Cabinets, Kewaunee
Carts
Two bay storage system to maximize storage capacity with divided trays, baskets and shelves.

  • Mobile cart
  • Procedure rooms
  • Cath lab, EP Lab, IR Lab, OR
  • Catheter hanging cabinet
  • Electronic keypad lock
  • Shelving
  • Supply storage

The cell storage system was specifically designed for use in the healthcare industry. High quality logistic system can be used in all departments of the hospitals. Because of the ability of its components, this system allows customers to find the optimal solution that answers to their specific needs while saving up to 30% of space in comparison to more traditional shelving systems.

Available with multiple lock options, including keyed lock, electronic keypad and electronic keypad with proximity card reader with two frequencies.

The cart is on four swivel locking casters for ease of movement. powder coated metal, 5" casters for easy maneuverability.

Ordering information: For more information on the different configurations, please contact your Avantor representative.

Delivery information: Carts include the trays, baskets and shelves that are shown with each part number and will be shipped inside of the cart.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty