Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EVERHUTCH CORECART Three-Bay Procedure Cabinets, Kewaunee

Supplier: Kewaunee

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MCT300-02-G3A MCT301-02-C3C MCT301-02-C3A MCT301-01-S3B MCT300-02-S3A MCT300-01-G3B MCT301-01-C3B MCT301-02-G3C MCT301-01-C3A MCT301-01-G3C MCT300-01-G3A MCT300-01-S3A MCT300-02-G3B MCT301-02-C3B MCT301-01-C3C MCT301-02-S3B
CT30101C3AEA 12900 USD InStock
CT30101C3A CT30102C3B CT30102C3A CT30101C3C CT30102C3C CT30101C3B CT30002S3A CT30001S3A CT30002G3A CT30001G3A CT30002G3B CT30001G3B CT30102S3B CT30101S3B CT30101G3C CT30102G3C
EVERHUTCH CORECART Three-Bay Procedure Cabinets, Kewaunee
Carts
Three bay storage system to maximize storage capacity with divided trays, baskets and shelves.

  • Mobile cart
  • Procedure rooms
  • Cath lab, EP Lab, IR Lab, OR
  • Catheter hanging cabinet
  • Electronic keypad lock
  • Shelving
  • Supply storage

The cell storage system was specifically designed for use in the healthcare industry. High quality logistic system can be used in all departments of the hospitals. Because of the ability of its components, this system allows customers to find the optimal solution that answers to their specific needs while saving up to 30% of space in comparison to more traditional shelving systems.

Available with multiple lock options, including keyed lock, electronic keypad and electronic keypad with proximity card reader with two frequencies.

The cart is on four swivel locking casters for ease of movement. powder coated metal, 5" casters for easy maneuverability.

Ordering information: For more information on the different configurations, please contact your VWR representative.

Delivery information: Carts include the trays, baskets and shelves that are shown with each part number and will be shipped inside of the cart.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty