Would you like to log in again?
VWR® Contour™ Apron Leg Assemblies
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CFA-2229-A CWA-1289-A CWA-2229-A CFA-1289-A CWA-2289-A CWA-1229-A CFA-2289-A CFA-1229-A
CWA-1289-AEA 289.04 USD InStock
CWA-1289-A CWA-2229-A CFA-1229-A CFA-2229-A CFA-2289-A CWA-1229-A CFA-1289-A CWA-2289-A
VWR® Contour™ Apron Leg Assemblies
Furniture Casework
A complete apron is used to create a knee space for a desk area or to create a free-standing table.
  • Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
  • No ragged edges
  • Light neutral color
Complete apron leg assembly. Apron components are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty