Would you like to log in again?
VWR® Contour™ Pedestal Legs
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CWA-2230-L CFA-2236-R CWA-2830-R CWA-2230-R CFA-2233-R CFA-2830-L CFA-2233-L CWA-2830-L CFA-2836-LA CWA-2236-R CFA-2230-RA CFA-2236-RA CWA-2836-R CFA-2836-L CFA-2236-LA CFA-2230-R CFA-2836-R CFA-2230-L CFA-2236-L CWA-2233-L CFA-2836-RA CWA-2233-R CFA-2830-R CWA-2236-L CWA-2836-L
CFA-2830-LEA 439 USD InStock
CFA-2830-L CFA-2230-RA CFA-2836-L CFA-2830-R CFA-2236-LA CFA-2836-R CWA-2230-L CWA-2233-L CFA-2836-RA CFA-2836-LA CFA-2236-RA CWA-2236-R CWA-2236-L CFA-2236-R CFA-2233-R CWA-2230-R CWA-2233-R CFA-2230-R CFA-2236-L CFA-2233-L CWA-2836-R CFA-2230-L CWA-2830-R CWA-2836-L CWA-2830-L
VWR® Contour™ Pedestal Legs
Furniture Casework
Contour Pedestal Legs are available in left- and right-specific models with varying depths and heights.
  • Available in a variety of heights and depths
Each pedestal leg adds 5.1 cm (2") to the overall length of the assembly to which it is attached.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty