Would you like to log in again?
VWR® Contour™ Apron
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CFA012218-A CFA022266-A CWA012248-A CFA012830-A CFA012248-A CWA012224-A CWA012242-A CWA022260-A CWA012236-A CFA012236-A CFA012224-A CWA022860-A CFA022872-A CFA022854-A CFA022848-A CWA022866-A CFA022260-A CFA022866-A CFA012230-A CFA022254-A CWA022254-A CFA012222-A CWA012230-A CFA012842-A CWA022272-A CFA012242-A CWA012830-A CWA022266-A CWA012218-A CFA022272-A CFA022860-A CWA022854-A
CWA012236-AEA 757 USD InStock
CWA012236-A CFA022848-A CFA022260-A CWA022854-A CWA012218-A CFA012830-A CWA022266-A CFA012236-A CWA022272-A CFA022872-A CFA012230-A CFA022266-A CFA012218-A CFA022854-A CWA012830-A CWA012224-A CWA022860-A CWA012248-A CFA022272-A CWA012242-A CWA022866-A CWA022254-A CFA012842-A CWA022260-A CFA012224-A CFA012248-A CFA022860-A CFA012242-A CFA012222-A CFA022254-A CFA022866-A CWA012230-A
VWR® Contour™ Apron
Furniture Casework
These aprons are used to create a knee space for a desk area or to create a free-standing table.
  • Steel construction
  • No ragged edges
Aprons are 4" high and made of steel.
Ordering information: Work surface and legs sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty