Would you like to log in again?
VWR® Contour™ ADA Apron Rails
Supplier: VWR
VWR®
CWA060336-A CFA060336-A
CWA060336-AEA 142.82 USD InStock
CWA060336-A CFA060336-A
VWR® Contour™ ADA Apron Rails
Furniture Casework
Apron Rails are 3" high (in lieu of standard 4" height) to comply with ADA requirements.
  • Steel
  • Only to be used between base cabinets
A complete apron is used to create a knee space for a desk area or to create a free-standing table (components sold separately).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty