Would you like to log in again?
VWR® Contour™ Knee Space Filler Panels
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CFA103148-A A10M2530-0000 CFA103142-A CWA103130-A CFA10M2524-00 A10M3136-00 CFA10M2542-00 CFA102548-A CWA102548-A CFA102524-A CWA103124-A CFA103136-A CFA10M2548-00 CFA102530-A CWA102536-A A10M3124-00 CWA103142-A CFA102542-A CWA103136-A CFA103130-A CFA102536-A CWA103148-A CWA102542-A CWA102530-A CFA103124-A
CWA103148-AEA 388.53 USD InStock
CWA103148-A CWA103124-A CWA102530-A CFA103142-A CFA10M3136-00 CWA102536-A CFA103124-A CFA103148-A CFA10M2542-00 CFA102530-A CFA10M2548-00 CFA10M3124-00 CWA103130-A CFA102536-A CFA10M2530-00 CWA103136-A CFA103130-A CWA102542-A CWA102548-A CFA102548-A CFA103136-A CFA10M2524-00 CFA102542-A CWA103142-A CFA102524-A
VWR® Contour™ Knee Space Filler Panels
Furniture Casework
Designed to close off the space between the floor and the bottom of sitting- or standing-height aprons
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Light neutral color
Panels are also used to conceal piping in furniture assemblies that include aprons. Two filler angles are included for mounting.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty