Would you like to log in again?
VWR® Contour™ Apron Support Rails
Supplier: VWR
VWR®
CFA802242-A CWA802248-A CFA802248-A CFA802230-A CWA802242-A CFA802236-A CWA802236-A CWA802230-A
CFA802242-AEA 476.21 USD InStock
CFA802242-A CFA802230-A CWA802236-A CWA802248-A CFA802248-A CFA802236-A CWA802230-A CWA802242-A
VWR® Contour™ Apron Support Rails
Furniture Casework
Worktop support frame.
  • Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
  • No ragged edges
  • Light neutral color
Apron support rails are available in a range of lengths.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty