Would you like to log in again?
VWR® Contour™ Mobile Drawer Unit With Pull Board
Supplier: VWR
VWR®
CFD-4002-B41 CWD-4002-B41
CWD-4002-B41EA 2510 USD InStock
CWD-4002-B41 CFD-4002-B41
VWR® Contour™ Mobile Drawer Unit With Pull Board
Furniture Storage Cabinets
Metal drawer unit in light neutral, on casters.
  • Drawer cabinets are furnished with counter weights to prevent tipping
  • Cabinets include an interlock system
  • All mobile cabinets have finished backs
Ordering information: Worksurface NOT included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty