Would you like to log in again?
VWR® Contour™ Suspended Cabinets
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CWE-4402-A CWD-4812-A CFD-4002-A CFD-5002-B CFC-5032-B CFE-4402-L CFC-4032-A CFC-4032-B CWD-4002-A CFE-4402-B CWC-5032-L CWE-4402-L CFC-4032-BL CFD-5852-B CFD-4812-A CFC-5032-L CFC-4032-L CFD-4852-B CWC-5032-A CWE-4402-B CFE-4402-A CWD-4852-B CFC-5032-A CFC-5032-BL CFE-4402-BL
CFD-4852-BEA 2020 USD InStock
CFD-4852-B CFC-4032-L CFD-4002-A CFC-5032-BL CFD-5852-B CFC-5032-A CFE-4402-BL CWD-4812-A CFC-5032-B CWD-4852-B CWE-4402-B CWE-4402-A CWC-5032-A CFE-4402-B CFC-5032-L CFE-4402-A CFE-4402-L CWE-4402-L CFC-4032-BL CFC-4032-A CFC-4032-B CWD-4002-A CWC-5032-L CFD-4812-A CFD-5002-B
VWR® Contour™ Suspended Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Cabinets are designed to hang beneath table frames. Light neutral color.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty