Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Contour™ Suspended Cabinets
Supplier: Avantor
VWR® Contour™
CFE-4404-L CWE-4404-A CFE-4404-A CWD-4854-A CWD-4004-A CFD-4854-A CWC-4034-A CFD-4814-A CFC-4034-A CFD-4004-A CWE-4404-L CWC-4034-L CWD-4814-A CFC-4034-L
CFD-4854-AEA 1891.6 USD InStock
CFD-4854-A CWC-4034-L CFE-4404-A CWE-4404-A CFC-4034-L CWD-4004-A CFE-4404-L CWE-4404-L CFD-4004-A CFC-4034-A CWD-4814-A CWC-4034-A CWD-4854-A CFD-4814-A
VWR® Contour™ Suspended Cabinets
Furniture Storage Cabinets
Powder-coated, steel, suspended cabinets mount directly to VWR® Lab tables.
  • Cabinets meet SEFA standards
  • Excellent chemical resistance
  • Soft close, positive catch doors
Suspended cabinets are available in a snow white or light neutral finish, with a variety of door and drawer configurations.
Ordering information: Mounting bar not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty