Would you like to log in again?
VWR® Contour™ Standing Height Cupboard and Drawer Base Cabinet
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CFE-2417-A E40M3724L03 CFE-2416-12 CFE-2408-A CWE-2418-A CFE-2407-A CFE-2402-L CFE-2402-12 CWE-2404-A CFE-2406-12 E40M3730-03 CFE-2404-A CWE-2402-L CWE-2404-L CWE-2408-A CFE-2404-L CWE-2415-A CWE-2402-A CFE-2404-12 CFE-2406-A CWE-2405-A CWE-2406-A CWE-2416-A CFE-2418-A CFE-2405-A CWE-2417-A C03M3718L03 CFE-2402-A CWE-2407-A CFE-2415-A CFE-2416-A
CFE-2416-12EA 2446.63 USD InStock
CFE-2416-12 CWE-2407-A CWE-2406-A CWE-2408-A CWE-2402-L CFE-2418-A CFE-2416-A CFE-2417-A CFE-2402-12 CFE-2404-12 CFE-2415-A CFE-2404-12L CFE-2406-12 CWE-2402-A CWE-2405-A CWE-2404-A CWE-2404-L CFE-2404-L CFE-2402-L CWE-2417-A CWE-2418-A CFE-2407-A CFE-2408-A CFE-2405-A CFE-2406-A CFE-2402-12L CFE-2405-12 CFE-2404-A CWE-2416-A CFE-2402-A CWE-2415-A
VWR® Contour™ Standing Height Cupboard and Drawer Base Cabinet
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface (not included) creates an overall height of 91.4 cm (36"). All cupboard cabinets have one full-width adjustable shelf.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty