Would you like to log in again?
VWR® Contour™ ADA Height Base Cabinets, Cupboard and Drawer Cabinets
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CWE-3402-A CFE-3402-A CWE-3402-L CFE-3456-L CFE-3456-A CFE-3402-L CWE-3456-A CWE-3456-L
CFE-3402-AEA 1699.51 USD InStock
CFE-3402-A CWE-3402-L CWE-3456-A CFE-3456-A CFE-3456-L CWE-3402-A CWE-3456-L CFE-3402-L
VWR® Contour™ ADA Height Base Cabinets, Cupboard and Drawer Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 83.8 cm (33"). All cupboard cabinets are supplied with one full-width adjustable shelf. Light neutral color.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty