Would you like to log in again?
VWR® Contour™ Standing Height Fume Hood Base Cabinet
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CWG-2085-A CWG-2086-A CFG-2086-A CFG-2088-A CFG-2085-A CFG-2084-0F2 CWG-2084-A CWG-2088-A CFG-2084-A CFG-2089-A
CFG-2088-AEA 2013.77 USD InStock
CFG-2088-A CFG-2086-A CFG-2085-A CFG-2084-0F2 CFG-2089-A CWG-2088-A CFG-2084-A CWG-2086-A CWG-2085-A CWG-2084-A
VWR® Contour™ Standing Height Fume Hood Base Cabinet
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). All cupboard cabinets have one full-width adjustable shelf.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty