Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Contour™ Acid Storage Fume Hood Base Cabinets
Supplier: VWR
VWR®
CFG-2808-V CFG-2804-VL CWG-2804-VL CFG-2806-V CWG-2804-V CFG-2804-V CWG-2805-V CFG-2805-V CWG-2806-V CWG-2808-V
CFG-2804-VLEA 2422.24 USD InStock
CFG-2804-VL CFG-2804-V CFG-2805-V CWG-2808-V CFG-2806-V CFG-2808-V CWG-2804-V CWG-2805-V CWG-2806-V CWG-2804-VL
VWR® Contour™ Acid Storage Fume Hood Base Cabinets
Furniture Storage Cabinets
VWR/VFH-06/19-8 Steel cabinets (35" high) acid storage fume hood base cabinets are specifically designed for the storage of corrosive chemicals.
  • 20" deep
  • Lined unit with vent
Available in either steel or wood. These cabinets are lined with a molded one piece linear low density polyethylene tub with coved corners and a 1" lip at the bottom front. The cabinet doors are lined with ⅛" sheet polyethylene and the doors are latched using a nylon roller catch. Each cabinet is furnished with a 1½" I.D. flexible polyolefin tube for venting to the fume hood above. (Requires a 2" hole in the hood work top.)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty