Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Contour™ Standing Height Base Cabinet for Acid Storage

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
CWG-2805-A CFG-2805-A CFG-2804-A CWG-2804-L CWG-2808-A CFG-2808-A CWG-2804-A CFG-2806-A CWG-2806-A CFG-2804-L
CFG-2805-AEA 2711.55 USD InStock
CFG-2805-A CFG-2806-A CFG-2808-A CWG-2804-L CWG-2808-A CFG-2804-L CWG-2804-A CWG-2805-A CWG-2806-A CFG-2804-A
VWR® Contour™ Standing Height Base Cabinet for Acid Storage
Furniture Casework
Heavy-gauge steel full-frame construction, specifically designed for the storage of corrosive chemicals.

  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Lined with a molded one piece linear LDPE tub with coved corners and a 1" lip
  • Cabinet doors are lined with 18" PE sheet
  • Doors are latched using a nylon roller catch

Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above

A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). Cupboard cabinets are supplied with one full-width adjustable shelf.

(Requires a 2" hole in the hood work-surface).

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty