Would you like to log in again?
VWR® Contour™ Standing Height Base Cabinet for Acid Storage
Supplier: VWR
VWR®
CWG-2805-A CFG-2805-A CFG-2804-A CWG-2804-L CWG-2808-A CFG-2808-A CWG-2804-A CFG-2806-A CWG-2806-A CFG-2804-L
CFG-2805-AEA 2711.55 USD InStock
CFG-2805-A CFG-2806-A CFG-2808-A CWG-2804-L CWG-2808-A CFG-2804-L CWG-2804-A CWG-2805-A CWG-2806-A CFG-2804-A
VWR® Contour™ Standing Height Base Cabinet for Acid Storage
Furniture Casework
Heavy-gauge steel full-frame construction, specifically designed for the storage of corrosive chemicals.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Lined with a molded one piece linear LDPE tub with coved corners and a 1" lip
  • Cabinet doors are lined with 18" PE sheet
  • Doors are latched using a nylon roller catch
Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above
A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). Cupboard cabinets are supplied with one full-width adjustable shelf.
(Requires a 2" hole in the hood work-surface).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty