Would you like to log in again?
VWR® Contour™ Base Cabinet Trim Parts
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CWL-1636-H CFL-0836-H CFL-0336-C CFL-0236-R CFL-0836-R CWL-0136-C CFL-0830-R CWL-0636-E CFL-1236-E CWL-1236-E CWL-0236-H CFL-0236-H CFL-0136-C CWL-0436-R CWL-0136-E CFL-2036-H CFL-0636-E CWL-0830-R CWL-0236-R CFL-1436-H CWL-0836-H CFL-0130-R CWL-2036-H CFL-0136-R CWL-0836-R CFL-0436-R CWL-0130-R CWL-0136-R CWL-1436-H CFL-1636-H CFL-0136-E
CWL-0136-REA 58.11 USD InStock
CWL-0136-R CWL-0130-R CWL-1436-H CWL-0136-E CWL-0136-C CWL-1236-E CFL-0136-E CFL-0130-R CFL-1436-H CWL-0836-H CWL-0830-R CFL-0136-R CFL-0636-E CWL-1636-H CFL-0836-R CWL-0836-R CFL-0236-H CWL-0436-R CFL-1636-H CWL-2036-H CFL-0136-C CFL-0436-R CFL-0336-C CFL-0236-R CFL-2036-H CWL-0236-R CWL-0636-E CFL-1236-E CFL-0830-R CWL-0236-H CFL-0836-H
VWR® Contour™ Base Cabinet Trim Parts
Furniture Casework
Rear scribe, corner, end scribe, and pipe chase trim parts are available.
  • Phosphate-coated steel with a baked, chemical resistant, powder-coat finish
  • Light neutral color
Base trim parts are available in varied heights.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty