Would you like to log in again?
VWR® Contour™ Full Height Storage Cabinets
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CWS25842236-A CFS25841348-A CFS12841636-A CWS00841636-A CFS12841336-A CFS12841348-A CWS25842248-A CFS11841636-A CFS25841648-A CFS12842236-A CFS25842248-A CWS12841648-A CFS25842236-A CFS00842248-A CFS12841648-A CWS25841348-A CFS00841648-A CWS12841636-A CFS12842248-A CFS00841636-A CFS00842236-A CFS25841636-A CWS25841636-A CWS00841648-A CWS25841648-A CFS00841336-A CFS11842248-A CFS20842248-A CFS00841348-A CFS25841336-A
CFS12842248-AEA 3789.73 USD InStock
CFS12842248-A CFS11841636-A CWS12841636-A CWS00841636-A CFS00842248-A CFS12841636-A CFS00841348-A CFS25842236-A CWS25842248-A CFS25841648-A CFS25841336-A CFS00841636-A CFS12841336-A CWS25841636-A CFS20842248-A CFS00841336-A CFS12842236-A CWS25841348-A CWS00841648-A CWS12841648-A CFS00842236-A CFS12841648-A CFS25841636-A CWS25842236-A CFS25841348-A CFS11842248-A CFS00841648-A CWS25841648-A CFS12841348-A CFS25842248-A
VWR® Contour™ Full Height Storage Cabinets
Furniture Casework
Standing height steel storage cabinets are 84" high and 123/4", 16", or 22" deep.
  • Five adjustable shelves
  • Light neutral color
Cabinets are phosphate-coated with a baked, chemical-resistant, powder-coat finish.
Caution: Full height cabinets may present a potential tipping over problem. Proper installation requires these cabinets to be anchored to a wall or other cabinets to improve stability and ensure smooth door movement.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty