Would you like to log in again?
VWR® Contour™ Wall Mounted Storage Cabinets with Sliding Plate Glass Doors
Supplier: VWR
VWR® Contour™
CFW10M3048 CWW10301348-A CFW10301336-A CFW10301348-A CWW10301336-A
CWW10301348-AEA 2060 USD InStock
CWW10301348-A CWW10301336-A CFW10301336-A CFW10M3048 CFW10301348-A
VWR® Contour™ Wall Mounted Storage Cabinets with Sliding Plate Glass Doors
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Wall cabinets come with two adjustable shelves.
Ordering information: Installation is required.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty