Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Contour™ Storage Cabinets with Glass Doors
Supplier: VWR
VWR® Contour™ VWR®
CFW50301324-L CWW50301324-L CFW50361324-L
CFW50301324-LEA 1236.11 USD InStock
CFW50301324-L CWW50301324-L CFW50361324-L
VWR® Contour™ Storage Cabinets with Glass Doors
Furniture Storage Cabinets
VWR Contour® tall units, wall-mounted units, or wall-mounted corner units.
  • Cold-rolled steel
  • Glazed glass doors
Wall-mounted units are available with sliding or swinging glass doors.
Certifications: Meets SEFA recommended practices.
Ordering information: Accessories not included.
Caution: Two person moving only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty