Would you like to log in again?
Ammonia, Vacu-vials Instrumental Kit, CHEMetrics
Supplier: CHEMetrics
K-1503
CXK-1503EA 61.26 USD InStock
CXK-1503
Ammonia, Vacu-vials Instrumental Kit, CHEMetrics
Environmental Test Kits
The ammonia test kits employing the well-established Nessler reagent to determine ammonia concentrations are applicable to drinking water, clean surface water, and good-quality nitrified wastewater effluent.
  • Analysis Time: 2 minutes
Why Test for Ammonia?
Ammonia is toxic to aquatic life, the aquaculture industry frequently measures and controls ammonia concentration to keep fish and shellfish alive. Regulatory requirements mandate ammonia monitoring for diverse industries to protect downstream aquatic environments. Examples include wastewater, landfill, acid mine drainage and animal feeding operations where ammonia is measured in effluent, leachate, and unintended discharges. Environmental groups often test water bodies for ammonia as part of their monitoring program to help them assess trends associated with other aquatic health indicators.
The VACUettes Auto Dilution Method: Hold the ampoule in a horizontal position while the capillary tip contacts the sample (Step 1). After the capillary fills, immerse it in a diluent (usually deionized water); snap the tip off the ampoule (Step 2). The sample and diluent are drawn into the ampoule where they mix with the reagent (Step 3). The resulting color change can then be compared with the flat or round comparator to quantify results (Step 4).
Seawater analysis requires A-1501 Stabilizer Solution (sold separately).
Ordering information: This kit requires the use of the CHEMetrics Direct-Readout Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Delivery information: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Stabilizer Solution, 25 mL sample cup, ampoule blank, and instructions.
Caution: Shelf life: Although the Nessler reagent is stable, its high alkali content attacks the glass ampoule. The resulting precipitate interferes with color comparison. The Vacu-vials ampoules have a two-month shelf life when stored in the dark and at room temperature. This shelf life can be extended by 18 months if the refill is refrigerated.
Contains mercury. Dispose according to local, state or federal laws.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty