Would you like to log in again?
Carbon Dioxide (Dissolved), Titrets Titration Cells, CHEMetrics
Supplier: CHEMetrics
Titrets®
K-1910
CXK-1910EA 67.13 USD InStock
CXK-1910
Carbon Dioxide (Dissolved), Titrets Titration Cells, CHEMetrics
Environmental Test Kits
Titrets® test kits use reverse titration to quantify concentrations in aqueous samples
  • Dependable, sealed single-use titrants
  • Fast, easy procedure — no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration anywhere
After snapping the ampoule tip, the sample is drawn into the ampoule in small doses (with the Titrettor device included in each kit that precisely controls the sample) (Step 1), until a color change signals that the equivalence point has been reached (Step 2). The titration is stopped at the end point and the ampoule is held upright. The liquid level will correspond to a printed scale on the ampoule's outer surface (Step 3).
Carbon dioxide ends up in surface and ground water as a result of animal respiration, decaying organic matter, and the decomposition of some minerals. Surface waters typically contain less than 10 ppm (mg/L) dissolved CO2, while ground waters, particularly if deep, may contain several hundred ppm (mg/L). Carbon dioxide concentration greatly impacts pH. In unpolluted waters, carbon dioxide is a major source of acidity.
Ordering information: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, Activator Solution, titrettor, 25 ml sample cup, and instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty