Would you like to log in again?
Iron (total & soluble), Vacu-vials Instrumental Kit, CHEMetrics
Supplier: CHEMetrics
K-6003
CXK-6003EA 60.74 USD InStock
CXK-6003
Iron (total & soluble), Vacu-vials Instrumental Kit, CHEMetrics
Environmental Test Kits
Ideal for low- to medium-range colorimetric analysis of water quality
  • Analysis Time: Soluble 1 minute; Total 5 minutes
The soluble iron content of surface waters rarely exceeds 1 mg/L, while ground waters often contain higher concentrations. The National Secondary Drinking Water Standard for iron is 0.3 mg/L, as iron concentrations in excess of 0.3 mg/L impart a foul taste and cause staining. High concentrations in surface waters can indicate the presence of industrial effluents or runoff.
Phenanthroline method: Ferrous iron reacts with 1,10-phenanthroline to form an orange-colored chelate. To determine total iron, thioglycolic acid solution is added to reduce ferric iron to the ferrous state. The reagent formulation minimizes interferences from various metals. Results are expressed as ppm (mg/L) Fe.
Ordering information: This kit requires the use of a CHEMetrics Direct-Readout Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Delivery information: Kit K-6003 comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Activator Solution, 25 ml sample cup, ampoule blank, and instructions.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty