Would you like to log in again?
COD Vials Kits, Chemetrics
Supplier: CHEMetrics
K-7375 K-7365 K-7360S K-7371S K-7351S K-7356 K-7360S K-7355 K-7375 K-7376 K-7366 K-7361S K-7350S K-7370S
CXK-7351SEA 62.32 USD InStock
CXK-7351S CACXK7355 CXK-7361S CXK-7350S CXK-7360S CXK-7371S CXK-7370S CACXK7375 CAK-7366 CACXK-7360 CXK7356 CXK7375 CXK7365 CXK7376
COD Vials Kits, Chemetrics
Environmental Test Kits
The kit provides two dichromate reactor digestion methods such as USEPA-accepted method, and a Mercury-free method for fast, easy, safe determinations of low, mid, and high-range COD levels in wastewater.
  • Leakproof reagent vials
  • Fast safe and easy COD determination
A fresh reagent ampoule blank must be prepared for each series of tests, therefore the number of samples that can be tested with each kit will vary.
Delivery information: Kits come in a cardboard box and contain everything needed to perform the tests (except distilled water) and instruction book.
Caution: These kits require the use of a Digestor Block along with a CHEMetrics Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16 mm round cell. All these instruments are sold separately.
Unless indicated as mercury-free, the kits contain mercury. Dispose according to local, state or federal laws.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty