Would you like to log in again?
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Supplier: CHEMetrics
CHEMets®
R-5510 K-5510
CXK5510EA 161.45 USD InStock
CXK5510 CXR-5510
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Environmental Test Kits
Hydrogen Peroxide CHEMets® used to to monitor sterilisation solutions in the packaging and sanitizing processes.
Hydrogen peroxide is a strong oxidizing agent used for treating of industrial effluents and domestic waste and serving as a disinfectant in aseptic packaging.
Delivery information: Kit includes refill, low and high range comparators, 25 ml sample cup, and instructions.
Packaging: Comes in a plastic case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty