Would you like to log in again?
Phosphate, ortho, CHEMets Visual Kit, CHEMetrics®
Supplier: CHEMetrics
CHEMets®
R-8515 R-8510 K-8530 K-8510 R-8510D R-8510
CXK-8510EA 131.07 USD InStock
CXK-8510 CXK-8530 CXR-8515 CACXR-8510 CXR-8510 CXR-8510D
Phosphate, ortho, CHEMets Visual Kit, CHEMetrics®
Environmental Test Kits
Ideal for low- to medium-range colorimetric analysis of water quality
CHEMets® test kits utilize self-filling ampules and vacuum pulls the sample up from a snap cup into the ampule when the tip is broken off. After tilting the ampule several times to mix the sample and reagent and waiting for the color to develop (usually 2 minutes or less), the analyst quantifies the result using the provided cylindrical comparator (for concentrations ≤1ppm) or flat comparator (for concentrations ≥1ppm).
Phosphorus occurs naturally in rock formations in the earth’s crust, usually as phosphate. High phosphate concentrations in surface waters may indicate fertilizer runoff, domestic waste discharge, or the presence of industrial effluents or detergents. Although phosphates from these sources are usually poly-phosphates or organically bound, all will degrade to ortho or reactive phosphates with time.
Phosphate measurement is used to control scale and corrosion inhibitor levels in boilers and cooling towers. Both methods Vanadomolybdophosphoric Acid and Stannous Chloride measure reactive phosphate, which will give a positive reaction prior to hydrolysis, and is usually termed ortho-phosphate.
Delivery information: Test kits contain 30 ampules, comparator(s), accessory solutions (when necessary), snap cup, and instructions. Refill packs contain 30 ampules.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty