Would you like to log in again?
Stainless Steel Drip Troughs, Durcon Laboratory Tops
Supplier: Durcon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SSDT24 SSDT30 SSDT36 SSDT18 SSDT48
DU-SSDT36EA 359.83 USD InStock
DU-SSDT36 DU-SSDT24 DU-SSDT30 DU-SSDT48 DU-SSDT18
Stainless Steel Drip Troughs, Durcon Laboratory Tops
Trays/Dishes/Pots Spillage Trays/Pans
Fabricated from Type 304 stainless steel with No
4 finish, drip troughs are furnished with an integral outlet for connection to a drain hose (included).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty