Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
POLYFIX® Bottle-Top Dispensers, MilliporeSigma
Supplier: MilliporeSigma
POLYFIX®
10107038-1 10107038-1 10107048-1 10107048-1
EM-10107048-1EA 804.02 USD InStock
CA10107038-1 CA10107048-1 EM-10107048-1 EM-10107038-1
POLYFIX® Bottle-Top Dispensers, MilliporeSigma
Bottle Top Dispensers
These dispensers are ideal for noncritical dispensing of light acids and aqueous solutions
Note: Do not use with solvents or concentrated acids.
They are made of polypropylene for durability and economy, and feature a calibrated glass cylinder. Volume setting system with additional fine adjustment is quick and easy-to-use. Units feature a scale rod on two sides for left- or right-handed use. The polypropylene valve block has a swivel base which allows dispenser to be fixed on the bottle in various positions. Unique neon green finger protection cap is ergonomically designed and aids in identification.
Ordering information: Supplied with a manufacturer's six-month warranty. Complete with an intake tube and a set of two polypropylene adapters (38 and 45mm). For additional adapter sizes, see EM-109050-1 series. Bottle not included.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty