Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Optifix® Adapters, MilliporeSigma
Supplier: MilliporeSigma
Optifix®
109129-1 109124-1 109219-1 109219-1 109245-1 109065-1 109128-1 109058-1 109040-1 109045-1 109038-1 109136-1 109128-1 109036-1 109036-1 109125-1 108059-1 109140-1 109245-1 109052-1 109140-1 109028-1 109124-1 109129-1 109050-1 109229-1 109025-1 109030-1 109138-1 109030-1 109050-1 109138-1 109136-1 109058-1 109028-1 109229-1 109040-1 109130-1 109038-1 109045-1 109065-1 109052-1 109125-1 109025-1 108059-1 109130-1
EM-109245-1EA 126.96 USD InStock
CAE19045-01 CA109038-1 CA109058-1 EM-109245-1 EM-109036-1 EM-109140-1 EM-109058-1 EM-109229-1 CAE19219-01 CAE19030-01 CAE19125-01 EM-109040-1 EM-109138-1 EM-109136-1 CAE19036-01 CAE19028-01 EM-109065-1 CAE19229-01 CAE19128-01 EM-109028-1 EM-109025-1 CAE19065-01 EM-109045-1 EM-109219-1 EMD-109038-1 CAE19052-01 CA109129-1 CAE19050-01 EM-109130-1 CAE19140-01 CAE19124-01 EM-108059-1 EM-109129-1 CAE19138-01 EM-109050-1 EM-109128-1 EM-109052-1 EM-109125-1 EM-109124-1 CAE19040-01 CAE19025-01 EM-109030-1 CAE19245-01 CAE19136-01 CAE19130-01 CAE18059-01
Optifix® Adapters, MilliporeSigma
Bottle Top Dispensers
For use with Optifix® and POLYFIX® dispensers (see EM-10108020-1 and EM-10107038-1 series)
Adapters fit standard screw-neck reagent bottles, and containers up to 200L drums. Sizes based on O.D. measurements.
Ordering information: Adapter set (EM-109050-1) includes four adapters: 24mm fits 4oz., 500mL, and 32oz. bottles; 28mm fits 4oz., 500mL, 16oz., 32oz., and 1L bottles; 38mm fits 1gal., 2.5L, 4L, 5gal., and 10gal. containers; and 45mm fits 10L containers. For additional adapter sizes, contact your VWR sales representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty