For use with Optifix® and POLYFIX® dispensers (see EM-10108020-1 and EM-10107038-1 series)
Adapters fit standard screw-neck reagent bottles, and containers up to 200L drums. Sizes based on O.D. measurements.
Ordering information: Adapter set (EM-109050-1) includes four adapters: 24mm fits 4oz., 500mL, and 32oz. bottles; 28mm fits 4oz., 500mL, 16oz., 32oz., and 1L bottles; 38mm fits 1gal., 2.5L, 4L, 5gal., and 10gal. containers; and 45mm fits 10L containers. For additional adapter sizes, contact your VWR sales representative.