Would you like to log in again?
OmniPur® PBS Liquid Concentrate 10X, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6504-20L 6504-20L 6505-4L 6505-4L
EM-6504EA 1354.98 USD InStock
CA6504 CA-EM6505 EM-6504 EM-6505
OmniPur® PBS Liquid Concentrate 10X, Calbiochem®, MilliporeSigma
Buffers Biological Buffers
Molecular biology grade 10X PBS liquid concentrate.
  • Clear, colorless liquid
  • DNase- and RNase-free
  • Intended for laboratory and manufacturing use only
  • Not for drug, food or household use
  • Freezable
Each liter of concentrate makes 10L of 1X Phosphate Buffered Saline (137mM Sodium Chloride, 2.7mM Potassium Chloride, 10mM Phosphate Buffer).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty