Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium hydrogen difluoride, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Ammonium bifluorideAmmonium hydrogen fluoride

AX1235-1 AX1235-5 AX1235-1 AX1235-5
EM-AX1235-1CS 597 USD InStock
CAAX1235-1 EM-AX1235-1 EM-AX1235-5 CAAX1235-5
Ammonium hydrogen difluoride, Supelco®
Ammonium hydrogen difluoride
Practical


Formula: NH₄HF₂
MW: 57.04 g/mol
Melting Pt: 127 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498
Order Now
Specification Test Results
Assay 96% min
Color White (typical)
Form Flakes

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty