Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia solution 28.0 - 30.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Synonyms: Ammonium hydroxide

M-Clarity™ quality level = MQ300

AX1303P-5 AX1303-6 AX1303P-5 AX1303-23 AX1303-3 AX1303-3 AX1303-75 AX1303-6
EM-AX1303P-5CS 1001 USD InStock
CAAX1303P-5 EM-AX1303P-5 EM-AX1303-75 EMD-AX1303-3 CAAX1303-3 EM-AX1303-6 EMAX1303-23 CAAX1303-6
Ammonia solution 28.0 - 30.0%, GR ACS, Supelco®
Ammonia solution


Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III
Order Now
Specification Test Results
Aluminum (Al) 0.3ppm max
Appearance passes test
Assay (as NH3) 28.0-30.0%
Calcium (Ca) 0.3ppm max
Carbon dioxide (CO2) 0.002% max
Chloride (Cl) 0.5ppm max
Chromium (Cr) 0.3ppm max
Copper (Cu) 0.1ppm max
Gold (Au) 0.3ppm max
Heavy metals (as Pb) 0.5ppm max
Iron (Fe) 0.2ppm max
Lead (Pb) 0.2ppm max
Magnesium (Mg) 0.3ppm max
Manganese (Mn) 0.2ppm max
Nickel (Ni) 0.1ppm max
Nitrate (NO3) 2ppm max
Phosphate (PO4) 2ppm max
Potassium (K) 0.3ppm max
Residue after ignition 0.002% max
Sodium (N/A) 0.3ppm max
Substances reducing permangaN/Ate To pass test
Sulfate (SO4) 2ppm max
Tin (Sn) 0.3ppm max
Titanium (Ti) 0.3ppm max
Zinc (Zn) 0.3ppm max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty