Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia solution 20 - 22%, OmniTrace Ultra™ for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Ammonium hydroxide

M-Clarity™ quality level = MQ100

AX1308-7 AX1308-7
EM-AX1308-7EA 411 USD InStock
CAAX1308-7 EM-AX1308-7
Ammonia solution 20 - 22%, OmniTrace Ultra™ for trace metal analysis, Supelco®
Ammonia solution
Clear liquid. For low level trace metal analysis. Double-distilled and packaged in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom conditions. Supplied in specially designed, preleached fluoropolymer resin bottles to guarantee product integrity. Certificate of Analysis supplied with each shipment.


Formula: NH₃
MW: 35.05 g/mol
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III
Order Now
Specification Test Results
Aluminum (Al) 20pg/g max
Antimony (Sb) 10pg/g max
Arsenic (As) 10pg/g max
Assay (as NH3) 20-22%
Barium (Ba) 10pg/g max
Beryllium (Be) 10pg/g max
Bismuth (Bi) 10pg/g max
Cadmium (Cd) 10pg/g max
Calcium (Ca) 10pg/g max
Cerium (Ce) 10pg/g max
Cesium (Cs) 10pg/g max
Chromium (Cr) 10pg/g max
Cobalt (Co) 10pg/g max
Copper (Cu) 10pg/g max
Dysprosium (Dy) 10pg/g max
Erbium (Er) 10pg/g max
Europium (Eu) 10pg/g max
Gadolinium (Gd) 10pg/g max
Gallium (Ga) 10pg/g max
Germanium (Ge) 10pg/g max
Gold (Au) 10pg/g max
Holmium (Ho) 10pg/g max
Indium (In) 10pg/g max
Iron (Fe) 10pg/g max
Lanthanum (La) 10pg/g max
Lead (Pb) 10pg/g max
Lithium (Li) 10pg/g max
Lutetium (Lu) 10pg/g max
Magnesium (Mg) 10pg/g max
Manganese (Mn) 10pg/g max
Mercury (Hg) 200pg/g max
Molybdenum (Mo) 10pg/g max
Neodymium (Nd) 10pg/g max
Nickel (Ni) 10pg/g max
Niobium (Nb) 10pg/g max
Potassium (K) 10pg/g max
Praseodymium (Pr) 10pg/g max
Rhodium (Rh) 10pg/g max
Rubidium (Rb) 10pg/g max
Samarium (Sm) 10pg/g max
Scandium (Sc) 10pg/g max
Silver (Ag) 10pg/g max
Sodium (N/A) 20pg/g max
Strontium (Sr) 10pg/g max
Tellurium (Te) 10pg/g max
Terbium (Tb) 10pg/g max
Thallium (Tl) 10pg/g max
Thorium (Th) 10pg/g max
Thulium (Tm) 10pg/g max
Tin (Sn) 10pg/g max
Titanium (Ti) 10pg/g max
Tungsten (W) 10pg/g max
Uranium (U) 10pg/g max
VaN/Adium (V) 10pg/g max
Ytterbium (Yb) 10pg/g max
Yttrium (Y) 10pg/g max
Zinc (Zn) 10pg/g max
Zirconium (Zr) 10pg/g max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty